Stampa…

N-(2-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride

Fornitore: Apollo Scientific

APOSPC408340-500MGEA 516 CHF
APOSPC408340-500MG
N-(2-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
N-(2-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
Formula: C₁₃H₁₉FN₂·ClH
Peso molecolare: 258,763 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD21098475
CAS Number: 1353959-43-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR