Stampa…

2-[(4-Fluoro-benzyl)-methyl-amino]-ethanol

Fornitore: Apollo Scientific

APOSPC408030-500MGEA 463 CHF
APOSPC408030-500MG
2-[(4-Fluoro-benzyl)-methyl-amino]-ethanol
2-[(4-Fluoro-benzyl)-methyl-amino]-ethanol
Formula: C₁₀H₁₄FNO
Peso molecolare: 183,223 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD14646020
CAS Number: 2248-70-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR