Stampa…

(5-Fluoro-pyridin-2-yl)-dimethyl-amine

Fornitore: Apollo Scientific

APOSPC408026-1GEA 1040 CHF
APOSPC408026-1G
(5-Fluoro-pyridin-2-yl)-dimethyl-amine
(5-Fluoro-pyridin-2-yl)-dimethyl-amine
Formula: C₇H₉FN₂
Peso molecolare: 140,158 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD24369772
CAS Number: 1420961-53-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR