Stampa…

Cerio (IV) solfato 0.1 mol/l (0.1 N) for 3S adapter technology soluzione volumetrica, Supelco®

Attenzione

Synonyms: Ceric disulfate

1.50693.1000EA 408 CHF
1.50693.1000
Cerio (IV) solfato 0.1 mol/l (0.1 N) for 3S adapter technology soluzione volumetrica, Supelco®
Cerio (IV) solfato

3S is an intelligent packaging solution protected with an originality seal. The cap provides information via Radio-frequency identification or RFID, enabling automated data transfer between the reagent and titration instrument. Intelligent communication for improved performance. 3S Reagents are compatible with Metrohm’s OMNIS titration platform through 3S Adapter Technology.


  • RFID-based identification of reagent—automated and error-free
  • Complete traceability of the analyses provided
  • Aspiration tube already integrated into the bottle
  • Unique opening procedure minimizes risk of contamination

Formula: Ce(SO₄)₂
Peso molecolare: 332,24 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00148852
CAS Number: 13590-82-4
UN: 3264
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR