Stampa…

Potassio disolfato ≥97.5% (by titrimetric analysis), Puriss. p.a., Fluka™

Pericolo

Synonyms: di-Potassio disolfato

60235-1KGEA 298.71 CHF
60235-1KG
Potassio disolfato ≥97.5% (by titrimetric analysis), Puriss. p.a., Fluka™
Potassio disolfato
Formula: K₂O₇S₂
Peso molecolare: 254,32 g/mol
Punto di fusione: 325 °C
Densità: 2,28 g/cm³ (25 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011385
CAS Number: 7790-62-7
EINECS: 232-216-8
UN: 2923
ADR: 8,II
Merck Index: 12,07834

Visualizza i codici

Risultati del test di specificazione

Assay 97.5 - 101.0 %
Silver (Ag) Max. 5 ppm
Aluminium (Al) Max. 5 ppm
Arsenic (As) Max. 5.0 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Thallium (Tl) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Phosphate (PO₄) Max. 10 ppm
Total N Max. 20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR