2-Hexylpyridine ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
A12772.14EA
1040
CHF
A12772.14
A12772.06
2-Hexylpyridine ≥98%
2-Hexylpyridine
Formula:
C₁₁H₁₇N Peso molecolare: 163,26 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00023552 CAS Number: 1129-69-7 EINECS: 214-454-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...