3-Chloro-2-methoxypyridine 95%
|
Attenzione
|
H27526.03EA
272
CHF
H27526.03
3-Chloro-2-methoxypyridine 95%
3-Chloro-2-methoxypyridine
Formula:
C₆H₆ClNO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03095227 CAS Number: 13472-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...