4-Bromo-2-fluoropyridine 95%
Fornitore: Thermo Scientific
|
Pericolo
|
H27657.06EA
671
CHF
H27657.06
4-Bromo-2-fluoropyridine 95%
4-Bromo-2-fluoropyridine
Formula:
C₅H₃BrFN Peso molecolare: 175,99 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04112504 CAS Number: 128071-98-7 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...