Ytterbium(III) fluoride, anidro ≥99.9% (REO, rare earth oxide basis)
|
Pericolo
|
41423.09EA
191
CHF
41423.09
41423.18
Ytterbium(III) fluoride, anidro ≥99.9% (REO, rare earth oxide basis)
Ytterbium(III) fluoride
For vacuum deposition
Formula:
F₃Yb Peso molecolare: 230,04 g/mol Punto di ebollizione: 2227 °C (1013 hPa) Punto di fusione: 1157 °C Densità: 8,2 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00049615 CAS Number: 13760-80-0 EINECS: 237-354-2 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...