Stampa…

Thulium(III) chloride 99.9% (trace metals basis)

Attenzione

ACRO316030010EA 70.4 CHF
ACRO316030010
Thulium(III) chloride 99.9% (trace metals basis)
Thulium(III) chloride
Formula: Cl₃Tm
Peso molecolare: 275,29 g/mol
Punto di fusione: 821 °C
MDL Number: MFCD00011283
CAS Number: 13537-18-3
EINECS: 236-904-9

Visualizza i codici

Risultati del test di specificazione

Appearance Off-white Powder
Total conc. of imp.as REO ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR