Stampa…

7-Aminoindole 97%

Fornitore: Thermo Scientific
Pericolo

L17794.MEEA 292 CHF
L17794.ME L17794.MC L17794.03
7-Aminoindole 97%
7-Aminoindole
Formula: C₈H₈N₂
Peso molecolare: 132,16 g/mol
Punto di fusione: 90…91 °C
MDL Number: MFCD02093960
CAS Number: 5192-04-1
EINECS: 000-000-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR