Stampa…

Indole-6-carboxylic acid

Fornitore: Molekula

MOLEM39891889EA 0 CHF
MOLEM39891889
Indole-6-carboxylic acid
Indole-6-carboxylic acid
Formula: C₉H₇NO₂
Peso molecolare: 161,16 g/mol
Punto di fusione: 254…257 °C
MDL Number: MFCD00210441
CAS Number: 1670-82-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR