5-Chloroindole
Fornitore: Molekula
MOLE25238892-5GEA
0
CHF
MOLE25238892-5G
5-Chloroindole
5-Chloroindole
Formula:
C₈H₆ClN Peso molecolare: 151,6 g/mol Punto di fusione: 71…72 °C |
MDL Number:
MFCD00005672 CAS Number: 17422-32-1 EINECS: 241-448-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...