Stampa…

6-Bromoindole

Fornitore: Molekula

MOLEM57350421EA 0 CHF
MOLEM57350421
6-Bromoindole
6-Bromoindole
Formula: C₈H₆BrN
Peso molecolare: 196,05 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00238550
CAS Number: 52415-29-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR