Stampa…

4-Bromoindole

Fornitore: Molekula

MOLE65921824-1GEA 0 CHF
MOLE65921824-1G
4-Bromoindole
4-Bromoindole
Formula: C₈H₆BrN
Peso molecolare: 196,05 g/mol
MDL Number: MFCD00671502
CAS Number: 52488-36-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR