Tetraoctylammonium bromide
Fornitore: Molekula
MOLE45634299-25GEA
0
CHF
MOLE45634299-25G
Tetraoctylammonium bromide
Tetraoctylammonium bromide
Formula:
C₃₂H₆₈BrN Peso molecolare: 546,8 g/mol Punto di fusione: ~ 96 °C Flash Pt: 100 °C |
MDL Number:
MFCD00011863 CAS Number: 14866-33-2 EINECS: 238-936-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...