Stampa…

(1-Nonyl)triphenylphosphonium bromide ≥98%

Attenzione

L02341.06EA 45 CHF
L02341.06
(1-Nonyl)triphenylphosphonium bromide ≥98%
(1-Nonyl)triphenylphosphonium bromide
Formula: H₃C(CH₂)₈P(C₆H₅)₃Br
Peso molecolare: 469,44 g/mol
Punto di fusione: 63…65 °C
MDL Number: MFCD00051875
CAS Number: 60902-45-6
EINECS: 262-521-1
https://ch.vwr.com/stibo/web/std.lang.all/96/55/2299655.jpg

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR