Stampa…

2,3,4,5,6-Pentamethylbenzyl alcohol ≥96%

B22574.03EA 26.5 CHF
B22574.03
2,3,4,5,6-Pentamethylbenzyl alcohol ≥96%
2,3,4,5,6-Pentamethylbenzyl alcohol
Formula: C₁₂H₁₈O
Peso molecolare: 178,27 g/mol
Punto di fusione: 158…160 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00014421
CAS Number: 484-66-2
EINECS: 207-609-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR