3-Cyanophenylboronic acid
Fornitore: Molekula
|
Attenzione
|
MOLEM46237605EA
0
CHF
MOLEM46237605
3-Cyanophenylboronic acid
3-Cyanophenylboronic acid
Formula:
C₇H₆BNO₂ Peso molecolare: 146,94 g/mol |
MDL Number:
MFCD01318967 CAS Number: 150255-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...