Stampa…

2-Ethylcaproic anhydride 95%

Pericolo

ACRO424790100EA 119 CHF
ACRO424790100
2-Ethylcaproic anhydride 95%
2-Ethylcaproic anhydride
Formula: C₁₆H₃₀O₃
Punto di ebollizione: 163 °C (16 mmHg)
Densità: 0,9909 g/cm³
Flash Pt: >212 °C
CAS Number: 36765-89-6
EINECS: 253-195-1
UN: 3265

Visualizza i codici

Risultati del test di specificazione

Appearance Clear colorless Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4335 to 1.4355 (20°C, 589 nm)
Titration by coupling ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR