Dibromoacetic acid
Fornitore: EHRENSTORFER GMBH
EHERC12216000EA
62
CHF
EHERC12216000
Dibromoacetic acid
Dibromoacetic acid
Formula:
Br₂CHCOOH Peso molecolare: 217,84 g/mol Punto di ebollizione: 238 °C (1013 hPa) Temperatura di stoccaggio: Ambiente |
CAS Number:
631-64-1 EINECS: 211-165-5 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...