D-(-)-Arginine
Fornitore: Molekula
|
Attenzione
|
MOLEM12156501EA
0
CHF
MOLEM12156501
D-(-)-Arginine
D-(-)-Arginine
Formula:
C₆H₁₄N₄O₂ Peso molecolare: 174,2 g/mol Punto di fusione: 212 °C |
MDL Number:
MFCD00063116 CAS Number: 157-06-2 EINECS: 205-866-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...