Stampa…

Dietilamina 20% (v/v) in acetonitrile, Novabiochem®

Fornitore: Merck
Pericolo


M-Clarity™ quality level = MQ200

MERC8.57010.0500EA 159 CHF
MERC8.57010.0500
Dietilamina 20% (v/v) in acetonitrile, Novabiochem®
Dietilamina

DEA/ACN Mix

Formula: (C₂H₅)₂NH
Peso molecolare: 73,14 g/mol
MDL Number: MFCD00009032
CAS Number: 109-89-7
UN: 2924
ADR: 3 (8),II
https://ch.vwr.com/stibo/web/std.lang.all/76/20/2297620.jpg

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR