Stampa…

D(+)-Raffinosio pentaidrato, Millipore®


M-Clarity™ quality level = MQ100

1.07419.1000EA 0 CHF
1.07419.1000
D(+)-Raffinosio pentaidrato, Millipore®
D(+)-Raffinosio pentaidrato
Formula: C₁₈H₃₂O₁₆·5H₂O
Peso molecolare: 594,52 g/mol
Punto di fusione: 80 °C
Densità: 1,516 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00071590
CAS Number: 17629-30-0
EINECS: 208-146-9
Merck Index: 12,08279

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR