Holmium ≥99.9%, in fogli, Thickness: 1.0 mm (0.04 in)
Fornitore: Thermo Scientific
10211.FFEA
586
CHF
10211.FF
Holmium ≥99.9%, in fogli, Thickness: 1.0 mm (0.04 in)
Holmium
Formula:
Ho Peso molecolare: 164,93 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00011049 CAS Number: 7440-60-0 EINECS: 231-169-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...