Stampa…

Glycyrrhizic acid monoammonium salt ≥95%, Millipore®

356780-5EA 267 CHF
356780-5
Glycyrrhizic acid monoammonium salt ≥95%, Millipore®
Glycyrrhizic acid monoammonium salt

A triterpenoid saponin isolated from liquorice roots that displays antiviral and anti-tumor activities. Inhibits replication, adsorption, and penetration of severe acute respiratory syndrome-associated coronavirus (SARS-CV), presumably by its ability to upregulate nitric oxide synthase activity.


  • White to off-white solid

Formula: C₄₂H₆₅NO₁₆
Peso molecolare: 839,97 g/mol
MDL Number: MFCD00167400
CAS Number: 53956-04-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR