Stampa…

Gomma arabica, milled powder NF

Attenzione

Synonyms: Acacia

SPCMGU117-45KGBLEA 0 CHF
SPCMGU117-45KGBL SPCMGU117-12KG SPCMGU117-2.5KG
Gomma arabica, milled powder NF
Gomma arabica

Melamine statement (available upon request)
BSE/TSE statement (available upon request)

Densità: 1,35 g/cm³ (20 °C)
Flash Pt: >250 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00081264
CAS Number: 9000-01-5
EINECS: 232-519-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR