Stampa…
Caffeina, anidro ≥98.5% USP
Pericolo
SPCMCA105-2.5KGEA 1610 CHF
SPCMCA105-2.5KG SPCMCA105-12KG SPCMCA105-50KGBL
Caffeina, anidro ≥98.5% USP
Caffeina
Caffeine, Anhydrous, USP is a central nervous system (CNS) stimulant of the methylxanthine class used in some migraine medications.
Formula: C₈H₁₀N₄O₂
Peso molecolare: 194,19 g/mol
Punto di fusione: 235 - 237,5 °C
MDL Number: MFCD00005758
CAS Number: 58-08-2
UN: 1544
ADR: 6.1,III
Merck Index: 13,01636
https://ch.vwr.com/stibo/web/std.lang.all/79/46/2297946.jpg
Visualizza i codici
Assay (Anhydrous Basis) 98.5 - 101.0 %
Water Determination max.0.5 %
Residue On Ignition max. 0.1 %
Heavy Metals (As Pb) max. 10 ppm
Organic Impurities To Pass Test
Learn more
About VWR
Mehr About VWR