Stampa…

Anti-PLAU Goat Polyclonal Antibody

Fornitore: United States Biological
USBIU2599-07NEA 1010 CHF
USBIU2599-07N
Anti-PLAU Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:8000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: PLAU (plasminogen activator, urokinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR