Stampa…

Anti-UCN2 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU2575-27AEA 1240 CHF
USBIU2575-27A
Anti-UCN2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: UCN2 (urocortin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR