Stampa…

Anti-UCK2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: United States Biological
USBIU2281-03-PEEA 1200 CHF
USBIU2281-03-PE
Anti-UCK2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:250; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: UCK2 (uridine-cytidine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR