Stampa…

Anti-UCK1 Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIU2280-91-BIOTEA 1200 CHF
USBIU2280-91-BIOT
Anti-UCK1 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: UCK1 (uridine-cytidine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR