Stampa…

Anti-UOX Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU2025-03AEA 1070 CHF
USBIU2025-03A
Anti-UOX Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: UOX (urate oxidase, pseudogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR