Stampa…

Anti-UBR2 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIU2020-02EA 774 CHF
USBIU2020-02
Anti-UBR2 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1: 32000

Type: Primary
Antigen: UBR2 (ubiquitin protein ligase E3 component n-recognin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR