Stampa…

Anti-USF2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: United States Biological
USBIU1740-04A-APCEA 1200 CHF
USBIU1740-04A-APC
Anti-USF2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: USF2 (upstream transcription factor 2, c-fos interacting)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR