Stampa…

Anti-USF2 Mouse Polyclonal Antibody

Fornitore: United States Biological
USBIU1740-01CEA 733 CHF
USBIU1740-01C
Anti-USF2 Mouse Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: USF2 (upstream transcription factor 2, c-fos interacting)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR