Stampa…

Anti-SLC25A27 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU1623-10-100EA 966 CHF
USBIU1623-10-100
Anti-SLC25A27 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: SLC25A27 (solute carrier family 25, member 27)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR