Stampa…

Anti-ULK1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: United States Biological
USBIU1500-70B-PEEA 1200 CHF
USBIU1500-70B-PE
Anti-ULK1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: ULK1 (unc-51 like autophagy activating kinase 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR