Stampa…

Anti-UBE2V1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU1075-01JEA 825 CHF
USBIU1075-01J
Anti-UBE2V1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: UBE2V1 (ubiquitin-conjugating enzyme E2 variant 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR