Stampa…

Anti-UCHL3 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU1005-01GEA 889 CHF
USBIU1005-01G
Anti-UCHL3 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry:10 ?g/ml

Type: Primary
Antigen: UCHL3 (ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR