Stampa…

Anti-Ubiquitin-activating enzyme E1L Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU1003-02EA 587 CHF
USBIU1003-02
Anti-Ubiquitin-activating enzyme E1L Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:5000-1:10000; Western Blot: 1:5000-1:10000

Type: Primary
Antigen: Ubiquitin-activating enzyme E1L
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR