Stampa…

Anti-UBA6 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU1002-01AEA 889 CHF
USBIU1002-01A
Anti-UBA6 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: UBA6 (ubiquitin-like modifier activating enzyme 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR