Stampa…

Anti-UBE4B Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: United States Biological
USBIU1001-60B-APCEA 1200 CHF
USBIU1001-60B-APC
Anti-UBE4B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: UBE4B (ubiquitination factor E4B)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR