Stampa…

Anti-UBE4A Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: United States Biological
USBIU1001-59-PEEA 1200 CHF
USBIU1001-59-PE
Anti-UBE4A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:200; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: UBE4A (ubiquitination factor E4A)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR