Stampa…

Anti-UBE2C Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornitore: United States Biological
USBIU1000-86B-FITCEA 1200 CHF
USBIU1000-86B-FITC
Anti-UBE2C Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: UBE2C (ubiquitin-conjugating enzyme E2C)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR