Stampa…
Anti-UBQLN3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: United States Biological
USBIU1000-20-PEEA 1200 CHF
USBIU1000-20-PE
Anti-UBQLN3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: UBQLN3 (ubiquilin 3)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR