Stampa…

Anti-UB Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU1000-07DEA 739 CHF
USBIU1000-07D
Anti-UB Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:1600

Type: Primary
Antigen: UB (ubiquitin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Zebrafish
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR