Stampa…

Anti-UB Rabbit Monoclonal Antibody [clone: 9H284]

Fornitore: United States Biological
USBIU1000-06PEA 1030 CHF
USBIU1000-06P
Anti-UB Rabbit Monoclonal Antibody [clone: 9H284]
Anticorpi
Recommended dilution: Flow Cytometry: 1:700; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500

Type: Primary
Antigen: UB (ubiquitin)
Clonality: monoclonal
Clone: 9H284
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rabbit, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR