Stampa…

Anti-Ubiquitin +1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIU1000-04TEA 889 CHF
USBIU1000-04T
Anti-Ubiquitin +1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:5000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Ubiquitin +1 (ubiquitin +1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR