Stampa…

Anti-Ubiquitin +1 Mouse Monoclonal Antibody [clone: 10K391]

Fornitore: United States Biological
USBIU1000-04PEA 672 CHF
USBIU1000-04P
Anti-Ubiquitin +1 Mouse Monoclonal Antibody [clone: 10K391]
Anticorpi
Recommended dilution: Western Blot: 1:500-1:1000;

Type: Primary
Antigen: Ubiquitin +1 (ubiquitin +1)
Clonality: monoclonal
Clone: 10K391
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR